Sterile distilled water and physiological solution
Sterile distilled water, 20×9 ml tubes, 10
Catalog # LIOF21097
Supplier: LIOFILCHEM
Sterile distilled water and physiological solution
Catalog # LIOF21097
Supplier: LIOFILCHEM
CAS Number:
Jump to:
Product Details & Documents
Sterile distilled water is generally used for the reconstitution of certain products or preparation of the initial suspension of a sample and further dilutions.
Store at 10 to 25 °C away from light. Do not use the product beyond its expiry date on the label or if product shows any evidence of contamination or any sign of deterioration.
Certifications: EN ISO 11133.
Caution: For laboratory and professional use only.
Specifications
Specifications
- VE:10
- Bezeichnung:Sterile distilled water, 20×9 ml tubes
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



