ContactUsLinkComponent
48 components: 10mg/l each of Al As Ba Be Bi B Cd Ca Ce Cs Cr Co Cu Dy Er Eu Gd Ga Ho In Fe La Pb Li Lu Mg Mn Nd Ni P K Pr Re Rb Sm Sc Se Na Sr Tb Tl Th Tm U V 1 * 100 mL
  CMPLRM007625L1
48 components: 10mg/l each of Al As Ba Be Bi B Cd Ca Ce Cs Cr Co Cu Dy Er Eu Gd Ga Ho In Fe La Pb Li Lu Mg Mn Nd Ni P K Pr Re Rb Sm Sc Se Na Sr Tb Tl Th Tm U V 1 * 100 mL
  CMPLRM007625L1
Frequently Bought Together