ContactUsLinkComponent
23 components: Ag 2.5mg/l Al 500mg/l As 25mg/l B 500mg/l Ba 500mg/l Be 25mg/l Cd 5mg/l Co 25mg/l Cr 25mg/l Cu 25mg/l Fe 500mg/l Li 25mg/l Mn 500mg/l Mo 25mg/l Ni 25mg/l Pb 25mg/l Sb 5mg/l Se 25mg/l Sr 500mg/l Tl 1 * 50 mL
Catalog # CMPLRM102331L05
23 components: Ag 2.5mg/l Al 500mg/l As 25mg/l B 500mg/l Ba 500mg/l Be 25mg/l Cd 5mg/l Co 25mg/l Cr 25mg/l Cu 25mg/l Fe 500mg/l Li 25mg/l Mn 500mg/l Mo 25mg/l Ni 25mg/l Pb 25mg/l Sb 5mg/l Se 25mg/l Sr 500mg/l Tl 1 * 50 mL
Catalog # CMPLRM102331L05
Frequently Bought Together