Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
CR2700 scanner, light grey, handled version, with B27 battery, W×D×H: 53,34×131,0×135,0 mm
Katalognr. BRDY176688
Anbieter: Brady
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
Katalognr. BRDY176688
Anbieter: Brady
Direkt zu
Product Details & Documents
Easily read wide and small barcodes with the ultra-fast CR2700 Handled barcode scanner. This aggressive, omnidirectional scanner eliminates repositioning and time consuming re-scans of barcodes.
- Uses B27 batteries with built-in power gauge
- IP 65 seal rating against dust and liquid ingress
- CodeShield® housing for disinfectant and chemical wipe down
- Unequalled performance reading on curved and shiny surfaces
- USB, Bluetooth®5 Low Energy (for enhanced data security, extended battery life, and less downtime)
Customisable visual, audible and haptic feedback with multiple programmable buttons. Trace any troubles with trackable service dates and battery statistics to help IT deploy and manage their scanner fleet.
Technische Daten
- Bezeichnung:CR2700 scanner, light grey, handled version, with B27 battery, W×D×H: 53,34×131,0×135,0 mm
- Seq:04
Accessories
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- BRDY145445Workstation lockout writer software appBBP®31, BBP®33, BBP®85, i3300Einloggen für Verfügbarkeit1 * 1 ST - SFr. 1’150.00
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- BRDY149445Brady Workstation Scan- und Druck-Software-SuiteBBP11, BBP12, BBP31, BBP33, BBP35, BBP37, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Colour and Cut, GlobalMark 2 Multicolour, i3300, i5100, i7100, i7500, IDXPERT, IP Series, J2000, J4000, J5000, LABXPERT, M410, M510, M610, M611, M710, PR Plus, S3100, THT 3" Core Printers, TLS 2200Einloggen für Verfügbarkeit1 * 1 ST - SFr. 1’150.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- BRDY195526Brady workstation CLP/GHS app via downloadEinloggen für Verfügbarkeit1 * 1 ST - SFr. 510.00
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- BRDY149187Brady workstation automation software suiteBBP11, BBP12, BBP30, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Multicolor, i3300, i5100, i7100, IP Series, J2000, J4000, J5000, M610, M611, M710, PR Plus, S3000, THT 76 mm Core Printers, Wraptor, Wraptor A6500Einloggen für Verfügbarkeit1 * 1 ST - SFr. 1’150.00