WEIGERTS IRON HEMATOXYLIN A 1 * 500 mL
Catalog # ELMI26044-05
Supplier: ELECTRON MICROSCOPY SCIENCES
WEIGERTS IRON HEMATOXYLIN A 1 * 500 mL
Catalog # ELMI26044-05
Supplier: ELECTRON MICROSCOPY SCIENCES
Frequently Bought Together
EPIKURE 3140 1 * 1 Fl
Supplier: MARKET SOURCE PART PROCESS
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond
EOSIN RAL 555 1 * 2,5 L
Supplier: RAL
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Nitrit VHR L, 500 ml
Supplier: Lovibond
Glycin (USP, BP, Ph. Eur.) reinst, Pharma-Qualität 1 * 1.000 g
Supplier: PanReac AppliChem
Allura red AC 80% (Farbstoffgehalt), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Direct Red 23, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Acid Red 183 30% (Farbstoffgehalt), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Oil Red EGN, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Bromocresol green-methyl red-Lösung gemischter Indikator, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Acid red 52, technische Qualität, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY