Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]MIX RD10003210.10 10X10ML AMPOULE 1 * 10 ST
Catalog # CMPLRD10003210.10
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
[EN]MIX RD10003210.10 10X10ML AMPOULE 1 * 10 ST
Catalog # CMPLRD10003210.10
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
[EN]VARIO CHLORINE FREE DPD F10 1 * 100 ST
Supplier: Lovibond
Acid red 27 ≥85-95% (Farbstoffgehalt), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Acid red 52, technische Qualität, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Oil Red EGN, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Bromocresol green-methyl red-Lösung gemischter Indikator, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN 1 * 20 L
Supplier: Hach
Acid Red 183 30% (Farbstoffgehalt), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
BLACK TISSUE MARKING DYE,59ML 1 * 1 ST
Supplier: Epredia
Test kits, photometric, AccuVac®
Supplier: Hach
Indigo 95% (Farbstoffgehalt), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY