Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]MIX RD10002910.10 10X10ML AMPOULE 1 * 10 ST
Catalog # CMPLRD10002910.10
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
[EN]MIX RD10002910.10 10X10ML AMPOULE 1 * 10 ST
Catalog # CMPLRD10002910.10
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
PHOSPHATE BUFFERED SALINE 1 * 10 L
Supplier: Avantor
Xylol, Isomerengemisch für die klinische Diagnostik 1 * 5 L
Supplier: PanReac AppliChem
Diluid III Diff für die Hämatologie, J.T.Baker®
Supplier: Avantor
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond