Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

NovaMop™ Self Wringing Mops

Self-wringing mop, 457 mm
Catalog # MICRQPNMSW-18
Supplier:   MICRONOVA
NovaMop™ Self Wringing Mops
Catalog # MICRQPNMSW-18
Supplier:  MICRONOVA
NovaMop™ Self Wringing Mops
NovaMop™ Self Wringing Mops

Self-wringing mop, 457 mm

Price
607.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

A flat head mop featuring a unique self-wringing mechanism that folds the mop head in half and draws upward for uniform wringing, every time. The 360° swivel joint allows full rotation of the mop head for use on floors, walls, and ceilings in the cleanroom.

  • Self wringing mop adapter with 360° swivel
  • One-step wringing process
  • Ideal, uniform wringing pressure
  • ACME thread adapter
  • Resistant to common cleaning chemicals and disinfectants
  • Autoclavable

The Micronova NovaMop™ Self Wringing Mop is designed for use with M-Zone® FlatMop™, SnapMop™, and PocketMop™ series flat head mops and connects directly to the STA series threaded handles, fixed length or extendible. Can also be used with SSU series stainless steel handles, with the addition of the SSA-3 adapter.

Accessories information: All components are ordered separately. For additional handle lengths, see Universal Mop Handles or Plastic Mop Handles from Micronova.

Specifications
  • Bezeichnung:
    Self-wringing mop
  • Länge:
    457 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.