TANGO-2 mini controller Control of 1 or 2 axes High position resolution: 819,200 microsteps / revolution Compact steel case Dimensions: 29 x 164 x 62 mm EMV suitable construction Solid steel case: robust, shielded plug connectors 1 * 1 ST
Katalognr. LEIM33080000
Anbieter: LEICA MICROSYSTEMS
Frequently Bought Together