AuftragsverfolgungKontakt
Guard Column Holders | Avantor
Guard Column Holders
Guard column holder (preparative, 'stand-alone') for 10×21,2 mm cartridges, ¹/₁₆" (10-32) thread on both sides
Katalognr. ISERAA13-000006
Anbieter:   ISERA
Guard Column Holders
Katalognr. ISERAA13-000006
Anbieter:  ISERA
Guard Column Holders
Guard column holder (preparative, 'stand-alone') for 10×21,2 mm cartridges, ¹/₁₆" (10-32) thread on both sides
Preis
SFr. 374.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Technische Daten
  • Bezeichnung:
    Guard column holder (preparative, 'stand-alone') for 10×21,2 mm cartridges, ¹/₁₆" (10-32) thread on both sides
  • Seq:
    06