Guard Column Holders
Guard column holder for direct coupling of 10 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
Katalognr. ISERAA13-000002
Anbieter: ISERA
Direkt zu
Product Details & Documents
Technische Daten
- Bezeichnung:Guard column holder for direct coupling of 10 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
- Seq:02