Guard Column Holders
Guard column holder for direct coupling of 5 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
Catalog # ISERAA13-000001
Supplier: ISERA
Jump to:
Product Details & Documents
Specifications
- Bezeichnung:Guard column holder for direct coupling of 5 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
- Seq:01