Liquid Dip Samplers
Liquid Dip sampler, Edelstahl 316L, 50 ml
Katalognr. SAMP5175A-50
Anbieter: Sampling Systems
Direkt zu
Product Details & Documents
The stainless steel liquid dip sampler is used to collect a wide variety of liquid samples from the surface.
- Made from high quality and chemically resistant 316L stainless steel
- Different sizes available
- Convenient hands-free emptying feature
Once the sample has been collected, the liquid dip sampler can simply be pushed against the container allowing for the liquid to be easily emptied.
Technische Daten
- Kapazität:50 ml
- Bezeichnung:Liquid Dip sampler
- Material:Edelstahl 316L
- Seq:01