Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Hand Scrapers
Hand scraper, white
  SAMP8336P-1
Hand Scrapers
  SAMP8336P-1
Hand Scrapers
Hand Scrapers
Hand scraper, white
31.70 SFr.
1 * 1 ST
These robust hand scrapers are perfect for scraping large areas.
  • Made from high quality PP
  • Robust and incorporates a large sturdy handle
  • Large 250 mm wide blade
  • Comes in three distinctive colours
The range of robust and sturdy hand scrapers from Sampling Systems are perfect for easily scraping large areas due to their large blade size and robust handle. The performance is further enhanced by the fact it is made out of PP giving it very good chemical resistance.
  • Bezeichnung:
    Hand scraper, white
  • Seq:
    03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.