[EN]5-METHOXYTRYPTAMINE 1 * 5 g
PROOTRCM271695-5G
Frequently Bought Together
[EN]TETRASUL 1 * 50 mg
: Merck
USP / NATIONAL FORMULARY FOR METHOD 467 1 * 1 mL
: EHRENSTORFER
TERT.-BUTYLHYDROQUINONE CERTIFIED 1 * 0,25 g
: EHRENSTORFER
in Acetonitrile 1 * 1 mL
: EHRENSTORFER
LOFEXIDINE HCL 1 * 1 mL
: Merck
n-Hexacosan, Supelco®
: Merck
AMARANTH CERTIFIED 1 * 0,25 g
: EHRENSTORFER
Gadolinium-Standardlösung, 1000 mg/L Gd in 5%iger Salpetersäure (aus Gd2O3) Specpure® Standard für die AAS
: Thermo Fisher Scientific
VISCOSITY STANDARD N4000 1 * 500 mL
: REAGECON
[EN]TETRABROMOBISPHENOL A-DIGLYCIDYL ETH 1 * 25 mg
: EHRENSTORFER
Diphenylzinndichlorid
: EHRENSTORFER
Buffer solutions, colour coded, Fluka™
: Honeywell Chemicals
THIOMETON 100MGM 1 * 100 mg
: EHRENSTORFER
TIOPRONIN CERTIFIED 1 * 0,1 g
: EHRENSTORFER
Indium Kalibrierstandard 1 * 1 ST
: Mettler - Toledo