SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
[EN]STANDARD AU 10MG/L IN HNO3 5% HCL 15 1 * 500 mL
CMPLRM087311L5.2
:
CUSTOM MADE CHEMICALS LAB
[EN]STANDARD AU 10MG/L IN HNO3 5% HCL 15 1 * 500 mL
CMPLRM087311L5.2
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Pufferlösungen in Beuteln, Certipur® , Supelco®
: Merck
pH Pufferlösungen, DuraCal
: HAMILTON BONADUZ
Leitfähigkeitsstandardlösungen
: VWR Chemicals
Pufferlösungen, gebrauchsfertig, Referenztemperatur 20 °C, Certipur®, Supelco®
: Merck
Kaliumchlorid 3 mol/l (3 N) in wässriger Lösung, Supelco®
: Merck
Pufferlösungen, Certipur®, farbcodiert, Supelco®
: Merck
Buffer solutions
: Mettler - Toledo
VWR® AVS TITRINORM Pufferlösungen, 20 °C
: VWR Chemicals
Buffer solutions as per DIN 19266-01: 2000
: WTW
Pufferlösung, Ready to Use
: WTW