20G PL PHOSPHONATE 2 (285 TESTS) 1 * 20 g
Catalog # WTIDPLPOW20PPHON2
Supplier: WATER ID PRIMELAB
Frequently Bought Together
UltraClear® für die Histologie, J.T.Baker®
Supplier: Avantor
Quaternäre Ammonium-Teststäbchen, MQuant®, Supelco®
Supplier: Merck
Ammoniaklösung ∼32%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acid Red 183 30% (Farbstoffgehalt), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
NEISSERS SOLUTION II FOR MICROSCOPY 1 * 100 mL
Supplier: Roth Carl
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Glycin (USP, BP, Ph. Eur.) reinst, Pharma-Qualität 1 * 1.000 g
Supplier: PanReac AppliChem
[EN]TARTRAZINE 2.5MG/ML IN DMSO 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
Ferrozine mono-sodium salt hydrate
Supplier: Hach
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond
EOSIN RAL 555 1 * 2,5 L
Supplier: RAL