20G PL DPD NITRITE (285 TESTS) 1 * 20 g
Katalognr. WTIDPLPOW20DPDNITR
Anbieter: WATER ID PRIMELAB
Frequently Bought Together
EOSIN RAL 555 1 * 1 L
Anbieter: RAL
Benzonase®
Anbieter: MERCK PRODUCTION CHEMICALS
65ML PL POLYACRYLATE 1 (65 TESTS) 1 * 65 mL
Anbieter: WATER ID PRIMELAB
Reagents for Phosphate Determination using Lovibond® MD photometers
Anbieter: Lovibond
PVP STABILIZED LUGOL SOLUTION 1 * 1 L
Anbieter: RAL
2 components: 100mg/l each of Rb Pr in HNO3 5% 1 * 50 mL
Anbieter: CUSTOM MADE CHEMICALS LAB
Kaliumhydroxid 5 mol/l (5 N) in wässriger Lösung
Anbieter: PROLABO CMP
SYR V6 TE 100UL 1 * 1 ST
Anbieter: MARKET SOURCE PART PROCESS
Reagents for alkalinity determination using Lovibond® MD photometers
Anbieter: Lovibond
IRIOTEC 8850 1 * 250 g
Anbieter: MARKET SOURCE PART PROCESS
N,N,N',N'-Tetramethyl-p-phenylendiamin Dihydrochlorid ≥95%, Pulver
Anbieter: SIGMA ALDRICH MICROSCOPY
[EN]MES MONOHYDRATE CGMP MANUFACTURED 1 * 12 kg
Anbieter: Spectrum Chemical