Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
10ML PERMANGANATE TIME TEST (285 TESTS) 1 * 10 mL
Catalog # WTIDPL10PTT
Supplier:
WATER ID PRIMELAB
Expand image
10ML PERMANGANATE TIME TEST (285 TESTS) 1 * 10 mL
Catalog # WTIDPL10PTT
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
UltraClear® für die Histologie, J.T.Baker®
Supplier: Avantor
Aluminium CRM, Standard Solutions, Supelco®
Supplier: Merck
Ameisensäure ≥99%, HiPerSolv CHROMANORM® für die LC-MS
Supplier: VWR Chemicals
Formalinlösung 10%, Q Path®, gepuffert
Supplier: VWR Chemicals
Quaternäre Ammonium-Teststäbchen, MQuant®, Supelco®
Supplier: Merck
Testsätze, photometrisch, CSB, LCK Küvetten-Tests
Supplier: Hach
Test kits, photometrisch, Nitrit, LCK-Küvetten-Tests
Supplier: Hach
Natriumhydroxid 6 mol/l (6 N) in wässriger Lösung für die biopharmazeutische Produktion
Supplier: VWR Chemicals