[EN]7 COMPONENTS EACH IN HNO3 10% 1 * 100 mL
Katalognr. CMPLRM065085L1
Anbieter: CUSTOM MADE CHEMICALS LAB
[EN]7 COMPONENTS EACH IN HNO3 10% 1 * 100 mL
Katalognr. CMPLRM065085L1
Anbieter: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
PHOSPHATE BUFFERED SALINE 1 * 10 L
Anbieter: Avantor
Reagents for ammonia determination using Lovibond® MD photometers
Anbieter: Lovibond
EPIKURE 3140 1 * 1 Fl
Anbieter: MARKET SOURCE PART PROCESS
[EN]TRICHLOROETHENE 2000MG/L IN METHANOL 1 * 5 Ampull
Anbieter: CUSTOM MADE CHEMICALS LAB
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥98% (durch HPLC, TLC), Sigma-Aldrich®
Anbieter: SIGMA ALDRICH MICROSCOPY
ALKALINITY TOTAL PHOTOMETR REAG. 500MG/L 1 * 50 ST
Anbieter: Palintest
Malachitgrün-Oxalat in Wasser, kristallin für die Mikroskopie, Sigma-Aldrich®
Anbieter: SIGMA ALDRICH MICROSCOPY
trans-4-Hydroxyzimtsäure ≥98% (durch HPLC), Sigma-Aldrich®
Anbieter: SIGMA ALDRICH MICROSCOPY
Diluid III Diff für die Hämatologie, J.T.Baker®
Anbieter: Avantor