Titanium(IV) oxide ≥99% USP
Poly bottle, 500 g
Katalognr. SPCMTI140-500GM
Anbieter: Spectrum Chemical
Titanium(IV) oxide ≥99% USP
Katalognr. SPCMTI140-500GM
Anbieter: Spectrum Chemical
CAS Nummer: 13463-67-7
Direkt zu
Product Details & Documents
Titanium Dioxide, USP is used as a food coloring. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Synonyme: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide
- CAS Nummer:13463-67-7
- Formel:TiO₂
- MDL-Nummer:MFCD00011269
- Molekulargewicht:79.88 g/mol
- Merck-Index:13,09549
- Siedepunkt:2900 °C (1013 hPa)
- Schmelzender Platingehalt:1855 °C
- Dichte:4,2 g/cm³ (20 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:236-675-5
Technische Daten
Ergebnisse des Spezifikationstests
- Assay:99% Min.
Technische Daten
- Packung:Poly bottle
- VE:500 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



