N-Boc-D-prolinal
Bottle, 100 g
Katalognr. APOSOR316011-100G
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Synonyme: 1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate, (R)-N-BOC-Prolinal, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, N-Boc-D-Prolinal, N-Boc-D-prolinal, N-(tert-Butoxycarbonyl)-D-prolinal, (R)-tert-butyl 2-formylpyrrolidine-1-carboxylate
- CAS Nummer:73365-02-3
- Formel:C₁₀H₁₇NO₃
- MDL-Nummer:MFCD01321389
- Molekulargewicht:199.25 g/mol
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
- VE:100 g



