Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

(±)-Epichlorhydrin 99%

Bottle, 25 g
Catalog # APOSOR922424-25G
Flammable
Health
Toxic
Corrosive
Supplier:   Apollo Scientific
(±)-Epichlorhydrin 99%
Catalog # APOSOR922424-25G
Supplier:  Apollo Scientific
CAS Number:  106-89-8
(±)-Epichlorhydrin 99%

Bottle, 25 g

CAS Number:  106-89-8
Price
79.70 SFr.
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane
  • CAS Number:106-89-8
  • Formula:C₃H₅ClO
  • MDL Number:MFCD00005132
  • Molecular Weight:92.52 g/mol
  • Boiling Pt:115…117 °C
  • Boiling Pt:115 - 117 °C
  • Melting Pt:–57 °C
  • Density:1,183
  • Flash Pt:28 °C
  • UN:2023
  • ADR:6.1,II
Specifications

Specifications

  • Packung:
    Bottle
  • VE:
    25 g

Specifications