MultiScreen 96-well Ultrafiltration Plate with Ultracel-10 membrane kD For use in the separation, concentration, desalting, preparation of proteins and other biomolecules. For research use only. Single use. 1 * 1 Pack.
MILFMAUF01005
: Merck
Frequently Bought Together