SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
[EN]MULTI-ION STANDARDS 7 COMPONENTS 100 1 * 100 mL
ACSD7344.K1.W.L1
:
CPACHEM
[EN]MULTI-ION STANDARDS 7 COMPONENTS 100 1 * 100 mL
ACSD7344.K1.W.L1
:
CPACHEM
Frequently Bought Together
Pufferlösungen, Certipur®, farbcodiert, Supelco®
: Merck
VWR® AVS TITRINORM Pufferlösungen, 20 °C
: VWR Chemicals
Leitfähigkeitsstandardlösungen
: VWR Chemicals
Pufferlösungen in Beuteln, Certipur® , Supelco®
: Merck
Kaliumchlorid 3 mol/l (3 N) in wässriger Lösung, Supelco®
: Merck
Pufferlösungen, gebrauchsfertig, Referenztemperatur 20 °C, Certipur®, Supelco®
: Merck
Pufferlösung, Ready to Use
: WTW
pH Standard Solutions, IUPAC, Certified
: Hach