Warning signs
Warning of pic-pack 307, Polyester, 25×22 mm
Katalognr. BRDY224134
Anbieter: Brady
Direkt zu
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
- Recommended by ISO 7010
- Colour: Yellow/Black
- Shape: Triangle
Technische Daten
- Bezeichnung:Warning of pic-pack 307
- Abmessungen:25×22 mm
- Material:Polyester
- Seq:02