Specifications:- Compatible Column ID: 0.53 mm (metal). Compatible Column OuterDiameter (OD): 0.75 mm. Ferrule Material: Stainless s teel. Ferrule Type: Straight. GC Component Type: Ferrule. Inner Diameter (ID): 0.8 mm. Manufacturer Compatibility: Ag 1 * 10 ST
Catalog # HEWLG2855-28506
Supplier: AGILENT HEWLETT PACKARD
Frequently Bought Together