Cole-Parmer® Essentials Replacement Cell Culturing Flasks
Glass cell culture flask only, 125 ml
Katalognr. TECN21837-36
Anbieter: Cole-Parmer
Cole-Parmer® Essentials Replacement Cell Culturing Flasks
Katalognr. TECN21837-36
Anbieter: Cole-Parmer
Direkt zu
Product Details & Documents
Enhance the life of your BS-200 series cell culture stirring systems.
- Constructed of borosilicate glass
The culture vessels incorporate a unique base design which, together with the bulb-ended stirrer, ensures the cells are lifted into suspension at the lowest possible speeds. This gentle stirring action promotes high cell yields by preventing cell shearing. Replacement flasks do not include caps or stirring mechanisms.
Technische Daten
- Kapazität:125 ml
- Bezeichnung:Glass cell culture flask only
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.