Fitting, LDPE, Straight, Hosebarb to Quick-Disconnect, 3/8" ID x 1/4" ID; 100/Pk
Katalognr. MFLX06290-10BLK
Anbieter: Avantor Fluid Handling
Über diesen Artikel
Provides easy single-handed disconnection.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Construction from low-density polyethylene (LDPE).
- Tüllengröße/Schlauch Innen-Ø (Port 1):3/8"
- Verbindungstyp:Quick-Disconnect to Barb
- Flow pattern:Straight
- Bezeichnung:Fitting, LDPE, Straight, Hosebarb to Quick-Disconnect, 3/8" ID x 1/4" ID; 100/Pk
- Seq:03
- Environmentally Preferable:
- Garantie:1 year
- VE:100
- Fitting material:LDPE
Frequently Bought Together