Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT WNT3A MOUSE (CHO CELLS-DERIVED, WITHOUT ANY TAG), 85-90% PURITY, PROVIDED WITH 10MG OF STABILIZER 1 * 10 µG
Catalog # MSPPAMS.RMW3AH-010
Supplier:
MARKET SOURCE PART PROCESS
Bild vergrößern
RECOMBINANT WNT3A MOUSE (CHO CELLS-DERIVED, WITHOUT ANY TAG), 85-90% PURITY, PROVIDED WITH 10MG OF STABILIZER 1 * 10 µG
Catalog # MSPPAMS.RMW3AH-010
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR® Premium Line, Serologische Pipetten
Supplier: VWR Collection
VWR®, Homogenisationsbeutel
Supplier: VWR Collection
Mikrotiterplatten, 96- und 384-Well, Nunc™
Supplier: Thermo Fisher Scientific
Serologische Pipetten, Stripette™
Supplier: Corning
VWR®, Kryofläschchen
Supplier: VWR Collection
VWR®, Transferpipetten, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Supplier: VWR Collection
Zellkulturkolben, Falcon®
Supplier: Corning
VWR®, Kryoboxen, Karton, 133×133 mm
Supplier: VWR Collection
VWR®, Transferpipetten, High Performance
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Zellkulturschalen, Falcon®
Supplier: Corning
VWR® Standard Line, Enghalsflaschen, mit Schraubverschluss
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF