Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
PCRmax® Alpha Cyclers | Avantor

PCRmax® Alpha Cyclers

Thermal cycler, quad 96-well capacity, 100/240 VAC, 50/60 Hz
Katalognr. TECN93945-12
Anbieter:   Cole-Parmer
PCRmax® Alpha Cyclers
Katalognr. TECN93945-12
Anbieter:  Cole-Parmer
PCRmax® Alpha Cyclers
PCRmax® Alpha Cyclers

Thermal cycler, quad 96-well capacity, 100/240 VAC, 50/60 Hz

Preis
SFr. 19’960.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Easily create, modify and access programs, saving time.

  • Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
  • Independently control each block for simplified temperature and space optimization
  • Software allows quick access to commonly used protocols

These compact thermal cyclers deliver reproducible results every run—with little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification.

A Program Wizard generates a protocol specific to your sequence in seconds—a quick way to optimise new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.

Technische Daten
  • Bezeichnung:
    Thermal cycler, quad 96-well capacity
  • Seq:
    04
  • Netzanschluss:
    100/240 VAC, 50/60 Hz
  • Cat. No.:
    TECN93945-12
  • Gradient:
    +
  • Schnittstellen:
    USB
  • B×T×H:
    700×330×535 mm
  • Temperaturbereich des Deckels:
    35...115 °C, or off
  • Blockhomogenität:
    ±0,3 at 55 °C
  • Max. Probenzahl:
    Four sets of 96×0,2 ml tubes or four 96-well plates
  • Temperaturgenauigkeit:
    ±0,25 at 50 °C
  • Heizrampenrate:
    3,4 °C/s
  • Temperaturbereich:
    10...100 °C
  • Gradiententemperaturbereich:
    1...29 °C
  • Programme:
    Max. 1000