PCRmax® Alpha Cyclers
Product Details & Documents
Easily create, modify and access programs, saving time.
- Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
- Independently control each block for simplified temperature and space optimization
- Software allows quick access to commonly used protocols
These compact thermal cyclers deliver reproducible results every run—with little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification.
A Program Wizard generates a protocol specific to your sequence in seconds—a quick way to optimise new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.
Dokumente
TDS 93945-AlphaCycler TDS Alpha Cycler
Technisches Datenblatt
Technische Daten
- Bezeichnung:Thermal cycler, dual 384-well and dual 96-well capacity
- Seq:02
- Netzanschluss:100/240 VAC, 50/60 Hz
- Cat. No.:TECN93945-06
- Gradient:+
- Schnittstellen:USB
- B×T×H:700×330×535 mm
- Temperaturbereich des Deckels:35...115 °C, or off
- Blockhomogenität:±0,3 at 55 °C
- Max. Probenzahl:Two 384-well plates, two sets of 96×0,2 ml tubes or two 96-well plates
- Temperaturgenauigkeit:±0,25 at 50 °C
- Heizrampenrate:3,4 °C/s
- Temperaturbereich:10...100 °C
- Gradiententemperaturbereich:1...29 °C
- Programme:Max. 1000
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



