PCRmax® Alpha Cyclers
Product Details & Documents
Easily create, modify and access programs, saving time.
- Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
- Independently control each block for simplified temperature and space optimization
- Software allows quick access to commonly used protocols
These compact thermal cyclers deliver reproducible results every run—with little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification.
A Program Wizard generates a protocol specific to your sequence in seconds—a quick way to optimise new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.
Dokumente
TDS 93945-AlphaCycler TDS Alpha Cycler
Technisches Datenblatt
Technische Daten
- Bezeichnung:Thermal cycler, triple 384-well and single 96-well capacity
- Seq:09
- Netzanschluss:100/240 VAC, 50/60 Hz
- Cat. No.:TECN93945-04
- Gradient:+
- Schnittstellen:USB
- B×T×H:700×330×535 mm
- Temperaturbereich des Deckels:35...115 °C, or off
- Blockhomogenität:±0,3 at 55 °C
- Max. Probenzahl:Three 384-well plates, one set of 96×0,2 ml tubes or one 96-well plate
- Temperaturgenauigkeit:±0,25 at 50 °C
- Heizrampenrate:3,4 °C/s
- Temperaturbereich:10...100 °C
- Gradiententemperaturbereich:1...29 °C
- Programme:Max. 1000
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



