Human Transferrin ELISA Kit
Human Transferrin ELISA kit, 96 tests
Katalognr. ABCAAB288174-1X96
Anbieter: Abcam
Direkt zu
Product Details & Documents
Human Transferrin ELISA kit is a single-wash 90-min Simplestep used to quantify Human Transferrin with a sensitivity of 4,396 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.
Technische Daten
Technische Daten
- Bezeichnung:Human Transferrin ELISA kit
- Tests pro Kit:96 tests
- Empfindlichkeit:4,396 pg/ml
- Reaktivität der primären Antikörper:Human
- Testdauer:One step
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB288174-1X96
- Lagertemperatur:4 ℃
- Format:Pre-coated
- Target protein:Transferrin
- Erkennungsmethode:Colorimetric
- Testtyp:Sandwich
- Probentyp:Cell culture media, Heparin Plasma, Citrate plasma, Serum, EDTA Plasma
- Erfassungsbereich:15,625 - 1000 pg/ml
- Regulatory status:RUO
Technische Daten
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



