TIG Welding Gauntlets
Product Details & Documents
The TIG Ultra Welding Gauntlet offers fantastic dexterity with a goat leather palm and excellent burn resistance due to the split leather backing. Designed for the professional TIG welder with aramid stitching for ultra durability.
- EN12477 Type A welding protection
- Premium quality leather welding gauntlet
- The ultimate in dexterity for welders
- Made from high quality durable goat leather
Ideal for welding and metal handling, these gloves feature reinforced Aramid stitching for maximum durability and burn protection.
Colour: Brown
Zertifizierung: EN ISO 21420: 2020 Dexterity 2, EN 388: 2016 + A1: 2018 3243X, EN 12477: 2001 Type A, EN 407: 2020 422X31. ANSI/ISEA 105: 2016 CUT Level A2. CE certified.
Dokumente
TDS A521 - TIG Welding Gauntlet
Technisches Datenblatt
Technische Daten
- Handschuhgröße:XXL
- Farbe:Braun
- Seq:03
- Cat. No.:PRTWA521BRRXXL
- Einweg / wiederverwendbar:wieder verwendbar
- Gepudert/ungepudert:Ungepudert
- Beidseitig tragbar / handspezifisch:Handspezifisch
- Latex / ohne Latex:Ohne Latex
- Material:Leather/polyester
- Sterility:Non sterile