Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
TIG Welding Gauntlets
L, Braun
Katalognr. PRTWA521BRRL
Anbieter:   Portwest
TIG Welding Gauntlets
Katalognr. PRTWA521BRRL
Anbieter:  Portwest
TIG Welding Gauntlets
TIG Welding Gauntlets
L, Braun
Preis
16.90 SFr.
Einloggen für Verfügbarkeit
1 * 1 PAIRE
Product Details & Documents
The TIG Ultra Welding Gauntlet offers fantastic dexterity with a goat leather palm and excellent burn resistance due to the split leather backing. Designed for the professional TIG welder with aramid stitching for ultra durability.
  • EN12477 Type A welding protection
  • Premium quality leather welding gauntlet
  • The ultimate in dexterity for welders
  • Made from high quality durable goat leather
Ideal for welding and metal handling, these gloves feature reinforced Aramid stitching for maximum durability and burn protection.
Colour: Brown
 : EN ISO 21420: 2020 Dexterity 2, EN 388: 2016 + A1: 2018 3243X, EN 12477: 2001 Type A, EN 407: 2020 422X31. ANSI/ISEA 105: 2016 CUT Level A2. CE certified.
Technische Daten
  • Handschuhgröße:
    L
  • Farbe:
    Braun
  • Seq:
    01
  • Cat. No.:
    PRTWA521BRRL
  • Einweg / wiederverwendbar:
    wieder verwendbar
  • Gepudert/ungepudert:
    Ungepudert
  • Beidseitig tragbar / handspezifisch:
    Handspezifisch
  • Latex / ohne Latex:
    Ohne Latex
  • Material:
    Leather/polyester
  • Sterility:
    Non sterile