Biosafety cabinets, Class II A2, Purifier®
Product Details & Documents
Purifier Logic+ Class II Type A2 Biosafety Cabinets are suitable for work with agents that require Biosafety Level 1, 2 or 3 containment.
- Electronically Commutated Motor (ECM)
- Constant Airflow Profile™ (CAP) Technology airflow monitoring system that communicates with the ECM to deliver the precise volume of air as required and automatically adjust as filters load without relying on airflow sensors
- Line of sight LCD information center with Filter Life Remaining bar graph, status line for alarm conditions and alerts to warn when filter life diminishes to 20% and 0%
Purifier Logic+ Class II Type A2 Biosafety Cabinets are suitable for work with agents that require Biosafety Level 1, 2 or 3 containment. Other appropriate applications include work with antioneoplastic drugs, genetic material, carcinogens, allergens and additional substances that generate hazardous airborne particulates.
Purifier Logic+ Class II, Type A2 Biosafety Cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection. When canopy-connected, the biosafety cabinets may be used for applications involving volatile toxic chemicals and radionuclides as an adjunct to microbiological research. These benchtop cabinets are available in 3', 4', 5' and 6' nominal widths.
: NSF International listed, ETL listed, CE conformity marking (230 volt models).
Technische Daten
- Bezeichnung:6' Purifier logic+ class II type A2 biosafety cabinet with accessory package
- Seq:06
- Cat. No.:LBCP302611031
- Stromzufuhr:230 V, 50/60 Hz, Schuko
- Innen B×T×H:1988,82×792,48×1567,18 mm
- Sash opening:254 mm
- Umweltfreundlichkeit:Energiesparend|Nachhaltige Materialien



