Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
Product Details & Documents
PIPETMAN®, recognized as the pipetting standard for its reliability and proven durability, is a fully adjustable, air-displacement pipette that uses disposable tips. PIPETMAN combines legendary accuracy, precision, and durability with easy pipetting and handling. The pipettes come in a large range of volumes with reduced pipetting forces for hours of comfortable pipetting.
- Legendary performance
- Easy pipetting
- Built-to-last and convenient
- Simple to maintain
Accurate and precise, PIPETMAN air-displacement pipettes are recognized as one of the world standard. They come in a large range of volumes, including single and multichannel models, allowing you to handle various sample types with confidence. PIPETMAN feels like PIPETMAN® Classic but with lower forces for enhanced pipetting habits with a standard pipette. Made of PVDF and stainless steel, PIPETMAN offers premium durability. Built-to-last, it is the economical solution for years of pipetting. Routine cleaning and maintenance are all that is required to keep PIPETMAN in top condition for years. The PIPETMAN design allows for easy access to user-serviceable parts, along with a fully autoclavable tip holder and ejector.
: Models equipped with a tip ejector are only sold with a stainless steel tip ejector. Plastic tip ejectors are available in the accessories.
Specifications
- Farbcode:Beige
- Volumen:2 - 20 µl
- Bezeichnung:P12x20, 3×10 measurements
- Seq:16
- Genauigkeit:±0,10 - ±0,40 µl
- Unpräzision:≤ 0,08 - ≤ 0,20 µl
- Empfohlene Spitzen:DL10 / DFL10 / D200 / D30
Accessories
Product Information
- Catalog #BezeichnungFarbeAvailabilityPrice
- 612-1816Rotierender Pipettenständer, CARROUSEL™, für 7 Gilson Pipetten (außer Pipetman Concept®)WeißLog in for availability1 * 1 ST - SFr. 183.00
Product Information
- Catalog #BezeichnungFarbeAvailabilityPrice
- GILSF161405Angewinkelter Pipettenständer, TRIO™, für 3 Gilson Pipetten (außer Pipetman Concept®)DurchscheinendLog in for availability1 * 1 ST - SFr. 136.00
Product Information
- Catalog #BezeichnungFarbeAvailabilityPrice
- GILSF161406Pipettenhalter, SINGLE™, für eine Gilson Pipette, kann auf jeder glatten, trockenen Oberfläche angebracht, oder mit Schrauben befestigt werdenWeißLog in for availability1 * 1 ST - SFr. 30.00
Product Information
- Catalog #BezeichnungAvailabilityPrice
Product Information
- Catalog #BezeichnungAvailabilityPrice
Product Information
- Catalog #BezeichnungAvailabilityPrice
Product Information
- Catalog #BezeichnungAvailabilityPrice
Product Information
- Catalog #BezeichnungAvailabilityPrice