Cadence Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Female Luer x Male Luer Lock
MFLX41507-01
Exceptional durability.
- Easy to disassemble for cleaning
- Stopcock handles turn 180°
- Autoclavable
These Cadence Science luer stopcocks are constructed of nickel-plated brass for enhanced durability. These stopcocks offer worry-free cleaning with easy disassembly to save valuable time.
- Flow pattern:One-Way
- Bezeichnung:Luer Stopcock Fitting, Nickel-Plated Brass, One-Way, Female Luer x Male Luer Lock; 1/Ea
- Environmentally Preferable:
- Wetted materials:Nickel-Plated Brass
- VE:1
- Inlet port:Female Luer
- Outlet port:Male Luer Lock
Frequently Bought Together