Barbed Fittings, Straight Connector, PFA, 3/8" ID
Katalognr. MFLX06383-97
Anbieter: Avantor Fluid Handling
Über diesen Artikel
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Chemical resistant
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- Tüllengröße/Schlauch Innen-Ø (Port 1):⅜"
- Tüllengröße/Schlauch Innen-Ø (Port 2):⅜"
- Verbindungstyp:Barbed Union
- Flow pattern:Straight
- Bezeichnung:Barbed Fittings, Straight Connector, PFA, ⅜" ID
- Environmentally Preferable:
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Garantie:1 year
- VE:1
- Fitting material:PFA
Frequently Bought Together