Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Cylindrical Barbed Cross Connectors, PVDF

Burkle Barbed Cross Connector, PVDF, 13-15 mm Tubing; 10/Pk, 15 mm, 15 mm, 15 mm, 15 mm, Barbed Union, PVDF, X
  MFLX06044-34
15 month warranty
Cylindrical Barbed Cross Connectors, PVDF
  MFLX06044-34
15 month warranty
Cylindrical Barbed Cross Connectors, PVDF
Cylindrical Barbed Cross Connectors, PVDF

Burkle Barbed Cross Connector, PVDF, 13-15 mm Tubing; 10/Pk, 15 mm, 15 mm, 15 mm, 15 mm, Barbed Union, PVDF, X

 
SFr. 41.30
   
1 * 10 ST
 

Safe and flexible connection for your most demanding applications.

  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable

The cylindrical barbed cross connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.

 
  • Tüllengröße/Schlauch Innen-Ø (Port 1):
    15 mm
  • Tüllengröße/Schlauch Innen-Ø (Port 2):
    15 mm
  • Verbindungstyp:
    Barbed Union
  • Flow pattern:
    X
  • Bezeichnung:
    Burkle Barbed Cross Connector, PVDF, 13-15 mm Tubing; 10/Pk
  • Barb size/tubing int.Ø (port 3):
    15 mm
  • Barb size/tubing int.Ø (port 4):
    15 mm
  • Fitting material:
    PVDF
  • Cat. No.:
    MFLX06044-34
  • Chemical resistance:
    Yes
  • Max. temperature:
    160 °C (320 °F)
  • Sterilisation:
    Autoclave
  • Garantie:
    15 Months
  • No. of ports:
    4