Masterflex® Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 3/32"; 25/PK
Katalognr. MFLX45508-32
Anbieter: Avantor Fluid Handling
CE certificate
Über diesen Artikel
Stellen Sie Ihre Prozessintegrität mit vollständiger Materialrückverfolgbarkeit sicher.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Verbindungstyp: Adapter Bulkhead-Innen-Luer auf Schlauchnippel, 1/4-28 UNF x 3/32"
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
- Tüllengröße/Schlauch Innen-Ø (Port 1):3/32"
- Verbindungstyp:Female Luer to Barb
- Flow pattern:Straight
- Bezeichnung:Bulkhead Innen-Luer, 1/4-28 UNF-Gewinde, auf 3/32“ Schlauchnippel, PP, 25/Pck
- Seq:02
- Environmentally Preferable:
- Connection type (port 1):Female luer
- Connection type (port 2):Hose barb
- No. of ports:2
- VE:25
- Fitting material:Polypropylene
Frequently Bought Together