Sunset yellow FCF
Glass, amber, 500 g
Catalog # SPCMFD155-500GM
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: Sodium 6-hydroxy-5-((4-sulphonatophenyl)diazenyl)naphthalene-2-sulphonate, Food yellow No. 5, FD and C Yellow No. 6
- CAS Number:2783-94-0
- Formula:C₁₆H₁₀N₂Na₂O₇S₂
- MDL Number:MFCD00036437
- Molecular Weight:452.36 g/mol
Specifications
Specifications
- Packung:Glass, amber
- VE:500 g



