Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro powder spatula, -, 150 mm, 3 mm
Catalog # RSGA80.532.003
Supplier:   RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
Catalog # RSGA80.532.003
Supplier:  RSG
Micro-Spatulas and Micro-Spoon, RSG Solingen
Micro-Spatulas and Micro-Spoon, RSG Solingen

Micro powder spatula, -, 150 mm, 3 mm

Price
SFr. 23.10
Log in for availability
1 * 1 ST
Product Details & Documents

Made from stainless steel 1.4301.

  • Rust-proof
  • Highly resistant against chemicals
  • With 6 edged grip for better handling
  • Autoclavable
Specifications
  • Steril:
    -
  • Bezeichnung:
    Micro powder spatula
  • Länge:
    150 mm
  • Seq:
    01
  • Klingenbreite:
    3 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.