ContactUsLinkComponent
Pullulan nominal Mw: 800000 Da, PDI: 1,50, 100 mg 1 * 100 mg
Katalognr. POSSPSS-PUL800K
Pullulan nominal Mw: 800000 Da, PDI: 1,50, 100 mg 1 * 100 mg
Katalognr. POSSPSS-PUL800K
Frequently Bought Together