Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
iFSP1 ≥98%

iFSP1 ≥98%

50 mg
Catalog # CAYM29483-50
Irritant
Supplier:   Cayman Chemical
iFSP1 ≥98%
Catalog # CAYM29483-50
Supplier:  Cayman Chemical
CAS Number:  150651-39-1
iFSP1 ≥98%

50 mg

CAS Number:  150651-39-1
Price
504.00 SFr.
Log in for availability
1 * 50 mg
Product Details & Documents

iFSP1 is an inhibitor of apoptosis inducing factor mitochondria-associated 2/ferroptosis suppressor protein 1 (AIFM2/FSP1)

Achtung! Bitte beachten: 98

Synonyms:  1-amino-3-(4-methylphenyl)-pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile, 1-Amino-3-p-tolyl-benzo[4,5]imidazo[1,2-a]pyridine-2,4-dicarbonitrile
  • CAS Number:150651-39-1
  • Formula:C₂₀H₁₃N₅
  • MDL Number:MFCD01572665
  • Molecular Weight:323.4 g/mol
  • Storage Temperature:Gefrierschrank
Specifications

Specifications

  • VE:
    50 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.