Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Echinocandin B

Echinocandin B

Glass Vial, 1 mg
Katalognr. CAYM25520-1
Anbieter:   Cayman Chemical
Echinocandin B
Katalognr. CAYM25520-1
Anbieter:  Cayman Chemical
CAS Nummer:  
Echinocandin B
Echinocandin B

Glass Vial, 1 mg

CAS Nummer:  
Preis
415.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 mg
Product Details & Documents

Echinocandin B inhibits 1,3-β glucan synthase; inhibits incorporation of radiolabeled glucose into alkali-insoluble glucan by S. cerevisiae spheroplasts; inhibits growth of C. albicans in vitro (MIC = 3.9 µg/ml after 2 days)

  • An echinocandin antifungal
  • Stability: ≥2 years

Documents

Technische Daten

Technische Daten

  • Packung:
    Glass Vial
  • VE:
    1 mg
  • Cat. No.:
    CAYM25520-1
  • Form:
    Powder
  • Purity:
    >95%
  • Lagerbedingungen:
    Freezer
  • Protein/Peptid-Name:
    Echinocandin B

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.