Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Zero pressure weather caps | Avantor
Zero pressure weather caps
Zero pressure weather caps,914 mm height,152 mm nominal diameter,9,1 kg shipping weight
Katalognr. LBCP4722200
Anbieter:   LABCONCO
Zero pressure weather caps
Katalognr. LBCP4722200
Anbieter:  LABCONCO
Zero pressure weather caps
Zero pressure weather caps
Zero pressure weather caps,914 mm height,152 mm nominal diameter,9,1 kg shipping weight
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Weathercaps are constructed of strong, corrosion-resistant PVC.
Caps fits atop standard PVC duct, and permit vertical discharge of effluent air above roofline for dispersion away from the building. Caps add little static pressure to the exhaust system.
Technische Daten
  • Bezeichnung:
    Zero pressure weather caps,914 mm height,152 mm nominal diameter,9,1 kg shipping weight
  • Seq:
    01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.