Inosin
Bottle, 10 g
Katalognr. APOSBII1001-10G
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Synonyme: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
- CAS Nummer:58-63-9
- Formel:C₁₀H₁₂N₄O₅
- MDL-Nummer:MFCD00066770
- Molekulargewicht:268.23 g/mol
- Schmelzender Platingehalt:212…213 °C
- Lagertemperatur:Raumtemperatur
Documents
Zertifikate
Suche nach ChargennummerTechnische Daten
Technische Daten
- Packung:Bottle
- VE:10 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



