Inosin
Bottle, 5 g
Catalog # APOSBII1001-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
- CAS Number:58-63-9
- Formula:C₁₀H₁₂N₄O₅
- MDL Number:MFCD00066770
- Molecular Weight:268.23 g/mol
- Melting Pt:212…213 °C
- Storage Temperature:Raumtemperatur
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Packung:Bottle
- VE:5 g



